missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-xL Antibody (OTI4A9), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70247AF594
This item is not returnable.
View return policy
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Bcl-xL | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BCL2L1 | |
| Recombinant fragment expressed in E.coli corresponding to amino acids 1-212 of human Bcl-xL. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI4A9 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
| 598.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction