missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-xL, Clone: BX006 + 2H12, Novus Biologicals™
Mouse Monoclonal Antibody
€ 264.00 - € 474.00
Specifications
| Antigen | Bcl-xL |
|---|---|
| Clone | BX006 + 2H12 |
| Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18156054
|
Novus Biologicals
NBP2-34290-0.02MG |
0.02 mg |
€ 264.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18166254
|
Novus Biologicals
NBP2-34290-0.1MG |
0.1 mg |
€ 474.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Bcl-xL | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| Q07817 | |
| 598 | |
| A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein (BX006 & 2H12) | |
| Primary | |
| Store at 4C. | |
| 27 kDa |
| BX006 + 2H12 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| BCL2L1 | |
| IgG | |
| Protein A purified | |
| Recognizes a protein of 27kDa, identified as the Bcl-X protein. This MAb shows no cross-reaction with Bcl-2 or Bax protein. Bcl-X has two isoforms, Bcl-XL (long), a 241 amino acid protein which suppresses cell death. And Bcl-XS (short), a 178 amino acid protein lacking a 63 amino acid domain which functions as a dominant inhibitor of Bcl-2. This MAb reacts with both Bcl-XS and Bcl-XL proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title