missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-XL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10783-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize Bcl-XL, Bcl-X (s) and Bcl-X(Beta).
BCL-XL is a part of the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. BCL-XL is located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential and controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. The longer isoform acts as an apoptotic inhibitor and the shorter form acts as an apoptotic activator.Specifications
| Bcl-XL | |
| Polyclonal | |
| Unconjugated | |
| BCL2L1 | |
| Apoptosis regulator Bcl X, BCL2L1, Bcl-XL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12048, 24888, 598 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P53563, Q07817, Q64373 | |
| BCL2L1 | |
| Bcl-XL Fusion Protein Ag1232 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction