missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL11B Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-262AF532
This item is not returnable.
View return policy
Description
BCL11B Polyclonal antibody specifically detects BCL11B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| BCL11B | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| B-cell CLL/lymphoma 11B, B-cell CLL/lymphoma 11B (zinc finger protein), B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein, B-cell lymphoma/leukaemia 11B, B-cell lymphoma/leukemia 11B, BCL-11B, BCL11B/TRDC fusion, COUP-TF-interacting protein 2, CTIP-2, CTIP2ZNF856B, hRit1, hRIT1-alpha, Radiation-induced tumor suppressor gene 1 protein, RIT1, zinc finger protein hRit1 alpha | |
| A synthetic peptide which maps to a region between residues 1 and 50 of human B-cell CLL/lymphoma 11B using the numbering given in Swiss-Prot entry Q9C0K0 (GeneID 64919). | |
| 0.1 mL | |
| Apoptosis, Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
| 64919 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction