missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL11B Mouse anti-Human, Clone:1F8G8, Abnova™
Mouse monoclonal antibody raised against recombinant human BCL11B.
Brand: Abnova MAB17526.100ug
This item is not returnable.
View return policy
Description
This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell malignancies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. [provided by RefSeq]Specifications
| BCL11B | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry (1:50-1:250) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. |
|
| BCL11B | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1F8G8 | |
| B-cell CLL/lymphoma 11B (zinc finger protein) | |
| In PBS (0.05% sodium azide) | |
| CTIP-2, CTIP2, RIT1, hRIT1-alpha | |
| Recombinant protein corresponding to amino acid 1-150 of human BCL11B from E. coli. | |
| 64919 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction