missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCL11B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16677274
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16677274 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16677274 Supplier Invitrogen™ Supplier No. PA5100794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BCL11B.

This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell malignancies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL11B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BCL11B
Gene Accession No. Q9C0K0
Gene Alias 9130430L19Rik; AI604821; ATL1; ATL1-alpha; ATL1-beta; ATL1-delta; ATL1-gamma; B cell CLL/lymphoma 11B; B cell leukemia/lymphoma 11B; B630002E05Rik; BAF chromatin remodeling complex subunit BCL11B; B-cell CLL/lymphoma 11B; B-cell CLL/lymphoma 11B (zinc finger protein); B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein; B-cell leukemia/lymphoma 11B; B-cell lymphoma/leukaemia 11B; B-cell lymphoma/leukemia 11B; B-cell lymphoma/leukemia 11B (predicted), 3 prime; B-cell lymphoma/leukemia 11B (predicted), 5 prime; BCL11B; BCL-11B; BCL11B, BAF complex component; BCL11B/TRDC fusion; COUP-TF interacting protein 2; COUP-TF-interacting protein 2; COUP-TF-interacting protein 2 long form; CTIP2; CTIP-2; hRit1; hRIT1-alpha; LOW QUALITY PROTEIN: B-cell lymphoma/leukemia 11B; mRit1; Radiation-induced tumor suppressor gene 1 protein; Rit1; zinc f; zinc finger protein hRit1 alpha; ZNF856B
Gene Symbols BCL11B
Host Species Rabbit
Immunogen A synthesized peptide derived from human BCL11B(Accession Q9C0K0), corresponding to amino acid residues G284-E334.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314423, 64919
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.