Learn More
BCL2 Mouse anti-Human, Clone: BCL2/796, Abnova™
Mouse monoclonal antibody raised against full length recombinant human BCL2.
Brand: Abnova MAB13419.100ug
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq]
Specifications
| BCL2 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ | |
| Bcl-2 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 596 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| BCL2/796 | |
| Mouse monoclonal antibody raised against full length recombinant human BCL2. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| BCL2 | |
| Recombinant protein corresponding to full length human BCL2. | |
| 100 μg | |
| Primary | |
| This monoclonal antibody shows no cross-reaction with Bcl-x or Bax protein. | |
| Store at 4°C. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.