Learn More
BCL2L1 & BAX Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand:  Abnova DI0068.1Set
Product Code. 16141094
  • € 677.00 / 1 set
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the BCL2L1 protein, and the other against the BAX protein for use in in situ Proximity Ligation Assay.
Specifications
Specifications
BCL2L1
Unconjugated
BCL-XL/S/BCL2L/BCLX/Bcl-X/DKFZp781P2092/bcl-xL/bcl-xS
1 Set
Primary
Antibody pair set content: 1. BCL2L1 rabbit purified polyclonal antibody (20 ug) 2. BAX mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
In situ PLA
BCL2-like 1
BCL2L1
RUO
598/581
Human
Ab Pair