Learn More
BCL2L1 & BAX Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DI0068.1Set
This item is not returnable.
View return policy
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the BCL2L1 protein, and the other against the BAX protein for use in in situ Proximity Ligation Assay.Specifications
| BCL2L1 | |
| Unconjugated | |
| BCL-XL/S/BCL2L/BCLX/Bcl-X/DKFZp781P2092/bcl-xL/bcl-xS | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. BCL2L1 rabbit purified polyclonal antibody (20 ug) 2. BAX mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| BCL2-like 1 | |
| BCL2L1 | |
| RUO | |
| 598/581 | |
| Human | |
| Ab Pair |