missing translation for 'onlineSavingsMsg'
Learn More

BCL2L10 Antibody (8A11G12) - BSA Free, Novus Biologicals™

Product Code. 18289371 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18289371 0.1 mL 0.1mL
18620497 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18289371 Supplier Novus Biologicals Supplier No. NBP261704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCL2L10 Monoclonal specifically detects BCL2L10 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL2L10
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 8A11G12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias Apoptosis regulator Bcl-B, apoptotic protein NrH, bcl2-L-10, BCL2-like 10 (apoptosis facilitator), bcl-2-like protein 10, BCLB, BCL-B, Boo, Diva, MGC129810, MGC129811
Gene Symbols BCL2L10
Host Species Mouse
Immunogen Purified recombinant fragment of human BCL2L10 (AA: 31-186) expressed in E. Coli.
Molecular Weight of Antigen 22 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10017
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.