missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2L11, Mouse anti-Human, Clone: 2F10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010018-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MAKQPSDVSSECDREGRQLQPAERPPQLRPGAPTSLQTEPQGNPEGNHGGEGDSCPHGSPQGPLAPPASPGPFATRSPLFIFMRRSSLLSRSSSGYFSFDSpecifications
| BCL2L11 | |
| Monoclonal | |
| Unconjugated | |
| BCL2-like 11 (apoptosis facilitator) | |
| BAM/BIM/BIM-alpha6/BIM-beta6/BIM-beta7/BOD/BimEL/BimL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2F10 | |
| In 1x PBS, pH 7.4 | |
| NM_138621 | |
| BCL2L11 | |
| BCL2L11 (NP_619527, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10018 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction