missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCL6 Monoclonal Antibody (BL6.02 (PG-B6p))
GREENER_CHOICE

Product Code. 11314463
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314463 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11314463 Supplier Invitrogen™ Supplier No. MA511493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-11493 targets BCL-6 in IHC (P) and Western blot applications and shows reactivity with Bovine, Human, Ovine, Porcine, Rabbit, Mouse and Rat samples. The MA5-11493 immunogen is recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein.

Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin's lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt's lymphomas and in nodular, lymphocyte predominant Hodgkin's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone BL6.02 (PG-B6p)
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene BCL6
Gene Accession No. P41182, P41183
Gene Alias B cell CLL/lymphoma 6; B cell leukemia/lymphoma 6; B-cell CLL/lymphoma 6; B-cell CLL/lymphoma 6 (zinc finger protein 51); B-cell leukemia/lymphoma 5; B-cell leukemia/lymphoma 6; B-cell lymphoma 5 protein; B-cell lymphoma 5 protein (BCL5); B-cell lymphoma 6 protein; B-cell lymphoma 6 protein homolog; B-cell lymphoma 6 protein transcript; BCL5; BCL-5; Bcl6; Bcl-6; BCL6 transcription repressor; BCL6A; cys his2 zinc finger transcription factor; cys-his2 zinc finger transcription factor; LAZ3; LAZ-3 protein; Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3); lymphoma-associated zinc finger gene on chromosome 3; protein LAZ-3; ZBTB27; zinc finger and BTB domain-containing protein 27; Zinc finger and BTB domain-containing protein 27 (ZBTB27); Zinc finger protein 51; Zinc Finger Protein 51 (ZNF51); zinc finger transcription factor BCL6S; ZNF51
Gene Symbols BCL6
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100156549, 100353941, 101110557, 12053, 303836, 539020, 604
Target Species Human, Mouse, Rat, Rabbit, Bovine, Sheep, Pig
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.