missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL6B Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00255877-B01P
This item is not returnable.
View return policy
Description
BCL6B Polyclonal antibody specifically detects BCL6B in Human samples. It is validated for Western Blot
Specifications
| BCL6B | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH59404.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 255877 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| BAZFBcl6-associated zinc finger protein, B-cell CLL/lymphoma 6 member B protein, B-cell CLL/lymphoma 6, member B, B-cell CLL/lymphoma 6, member B (zinc finger protein), ZBTB28, Zinc finger protein 62ZNF62 | |
| BCL6B (AAH59404.1, 1 a.a. - 480 a.a.) full-length human protein. MGSPAAPEGALGYVREFTRHSSDVLGNLNELRLRGILTDVTLLVGGQPLRAHKAVLIACSGFFYSIFRGRAGVGVDVLSLPGGPEARGFAPLLDFMYTSRLRLSPATAPAVLAAATYLQMEHVVQACHRFIQASYEPLGISLRPLEAEPPTPPTAPPPGSPRRSEGHPDPPTESRSCSQGPPSPASPDPKACNWKKYKYIVLNSQASQAGSLVGERSSGQPCPQARLPSGDEASSSSSSSSSSSSEEGPIPGPQSRLSPTAATVQFKCGAPASTPYLLTSQAQDTSGSPSERARPLPGSEFFSCQNSEAVAGCSSGLDSLVPGDEDKPYKCQLCRSSFRYKGNLASHRTVHTGEKPYHCSICGARFNRPANLKTHSRIHSGEKPYKCETCGSRFVQVAHLRAHVLIHTGEKPYPCPTCGTRFRHLQTLKSHVRIHTGEKPYHCDPCGLHFRHKSQLRLHLRQKHGAATNTKVHYHILGGP | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction