missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL7B Monoclonal Antibody (OTI2D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527163
This item is not returnable.
View return policy
Description
BCL7B Monoclonal Antibody for Western Blot
Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1. Involved in cell cycle progression, maintenance of the nuclear structure and stem cell differentiation. May play a role in lung tumor development or progression.
Specifications
| BCL7B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BQE9 | |
| BCL7B | |
| Full length human recombinant protein of BCL7B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2D9 | |
| Unconjugated | |
| BCL7B | |
| B cell CLL/lymphoma 7B; B-cell CLL/lymphoma 7 protein family member B; B-cell CLL/lymphoma 7B; BCL tumor suppressor 7B; Bcl7b; Hom s 3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9275 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction