missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL7B Monoclonal Antibody (OTI7H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809577
This item is not returnable.
View return policy
Description
Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1. Involved in cell cycle progression, maintece of the nuclear structure and stem cell differentiation. May play a role in lung tumor development or progression.Specifications
| BCL7B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQE9 | |
| BCL7B | |
| Full length human recombit protein of human BCL7B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7H1 | |
| Unconjugated | |
| BCL7B | |
| BCL tumor suppressor 7B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9275 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction