missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL7B Monoclonal Antibody (OTI7H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809577S
This item is not returnable.
View return policy
Description
BCL7B Monoclonal antibody specifically detects BCL7B in Human samples. It is validated for Western BlotSpecifications
| BCL7B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQE9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9275 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7H1 | |
| Unconjugated | |
| BCL7B | |
| B cell CLL/lymphoma 7B; B-cell CLL/lymphoma 7 protein family member B; B-cell CLL/lymphoma 7B; BCL tumor suppressor 7B; Bcl7b; Hom s 3 | |
| Full length human recombinant protein of human BCL7B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction