missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCMA Monoclonal Antibody (1D12G9)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531673

Product Code. 16365104

  • € 516.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-31673 has been tested in indirect ELISA.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is preferentially expressed in mature B lymphocytes, and may be important for B cell development and autoimmune response. This receptor has been shown to specifically bind to the tumor necrosis factor (ligand) superfamily, member 13b (TNFSF13B/TALL-1/BAFF), and to lead to NF-kappaB and MAPK8/JNK activation. This receptor also binds to various TRAF family members, and thus may transduce signals for cell survival and proliferation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BCMA
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q02223
Tnfrsf17
Purified recombinant fragment of human BCMA (AA: QMAGQCSQNEYFDSLc) expressed in E. coli (aa 3-17).
100 μL
Primary
Human
Antibody
IgG1
ELISA, Flow Cytometry, Western Blot
1D12G9
Unconjugated
Tnfrsf17
B cell maturation antigen; B-cell maturation factor; B-cell maturation protein; BCM; BCMA; CD269; TNF receptor superfamily member 17; Tnfrsf13; Tnfrsf13a; Tnfrsf17; Tumor necrosis factor receptor superfamily member 17; tumor necrosis factor receptor superfamily, member 17
Mouse
Protein G
RUO
608
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.