Learn More
BCMA Neutralizing Mouse, Unconjugated, Monoclonal Antibody, recommended for neutralizing assay, AcroBiosystems
Description
BCMA Neutralizing Monoclonal antibody specifically detects BCMA Neutralizing, recommended for neutralizing assay.
Specifications
Specifications
| Antigen | BCMA Neutralizing |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Human (HEK293) |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | -20°C |
| Isotype | IgG1 |