missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCMA/TNFRSF17 Antibody (rCD269/8507), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20813CL1
This item is not returnable.
View return policy
Description
BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| BCMA/TNFRSF17 | |
| Monoclonal | |
| CoraFluor 1 | |
| B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 | |
| Recombinant human BCMA/TNFRSF17 protein fragment (around aa 1-184) (exact sequence is proprietary) | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction | |
| 608 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| rCD269/8507 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction