Learn More
BCMA/TNFRSF17 Antibody (rCD269/8507), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20418-100ug
 View more versions of this product
Code produit. 18672105
  • € 501.00 / 100µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
BCMA/TNFRSF17
Monoclonal
0.2 mg/ml
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Recombinant human BCMA/TNFRSF17 protein fragment (around aa 1-184) (exact sequence is proprietary)
100 μg
Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction
608
Store at 4&drg;C. Do not freeze.
IgG1 κ
ELISA, Immunohistochemistry (Paraffin)
rCD269/8507
Unconjugated
10mM PBS with 0.05% BSA