missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCMA/TNFRSF17, Rat anti-Human, PE, Clone: Vicky-1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-97656-0.1MG
This item is not returnable.
View return policy
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.
Specifications
| BCMA/TNFRSF17 | |
| Monoclonal | |
| PE | |
| PBS | |
| TNFRSF17 | |
| Recombinant human BCMA extracellular domain (aa 2-54). | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| Vicky-1 | |
| Flow Cytometry 1:100 | |
| B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 608 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction