missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCMO1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92267-0.1ml
This item is not returnable.
View return policy
Description
BCMO1 Polyclonal antibody specifically detects BCMO1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BCMO1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BCDO1, BCDOEC 1.14.99.36, BCMO, BCO, BCO1, beta, beta-carotene 15, 15'-dioxygenase 1, beta-carotene 15,15'-monooxygenase, beta-carotene 15,15'-monooxygenase 1, Beta-carotene dioxygenase 1, FLJ10730 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 268-547 of human BCMO1 (NP_059125.2). MSWASCLAFHREEKTYIHIIDQRTRQPVQTKFYTDAMVVFHHVNAYEEDGCIVFDVIAYEDNSLYQLFYLANLNQDFKENSRLTSVPTLRRFAVPLHVDKNAEVGTNLIKVASTTATALKEEDGQVYCQPEFLYEGLELPRVNYAHNGKQYRYVFATGVQWSPIPTKIIKYDILTKSSLKWREDDCWPAEPLFVPAPGAKDEDDGVILSAIVSTDPQKLPFLLILDAKSFTELARASVDVDMHMDLHGLFITDMDWDTKKQAASEEQRDRASDCHGAPLT | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 53630 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction