missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCOR Antibody (OTI4F6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71973PE
This item is not returnable.
View return policy
Description
BCOR Monoclonal antibody specifically detects BCOR in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BCOR | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 54880 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot | |
| OTI4F6 | |
| Western Blot | |
| ANOP2, BCL6 corepressor, BCL-6 corepressor, BCL6 co-repressor, BCL-6 interacting corepressor, BCoR, FLJ20285, FLJ38041, KIAA1575MGC71031, MAA2, MCOPS2, MGC131961 | |
| Human recombinant protein fragment corresponding to amino acids 384-668 of human BCOR(NP_060215) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction