missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCR/ABL Antibody (4C12), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2665JF646
This item is not returnable.
View return policy
Description
BCR/ABL Monoclonal specifically detects BCR/ABL in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| BCR/ABL | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BCR | |
| BCR/ABL peptide | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 4C12 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ABL, ALL, BCR ABL fusion protein, bcr/abl, BCR/ABL fusion protein, Bcr/c abl oncogene protein, BCR-ABL fusion protein, BCRe18/ABL1e3 fusion protein, breakpoint cluster region protein, c-ABL, CML, D22S662, JTK7, p150, PHL | |
| Mouse | |
| Protein A purified | |
| Protein Kinase, Signal Transduction | |
| 613 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction