missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bcr Polyclonal Antibody
GREENER_CHOICE

Product Code. 17741424
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17741424 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17741424 Supplier Invitrogen™ Supplier No. PA5120023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, Jurkat, HeLa, Mouse brain, Rat brain. The target is usually found in the following locations: Cell junction, postsynaptic cell membrane, postsynaptic density, synapse. Immunogen sequence: MVDPVGFAEA WKAQFPDSEP PRMELRSVGD IEQELERCKA SIRRLEQEVN QERFRMIYLQ TLLAKEKKSY DRQRWGFRRA AQAPDGASEP RASASRPQPA PADGADPPPA EEPEARPDGE GSPGKARPGT ARRPGAAASG ERDDRGPPAS VAALRSNFER IRKGHGQPGA DAEKPFYVNV EFHHERGLVK VNDKEVSDRI SSLGSQAMQM ERKKSQHGAG SSVGDASRPP YRGRSSESSC GVDGDYEDAE LNPRFLKDNL IDANGGSRPP WPPLEYQPYQ SIYVGGMMEG EGKGPLLRSQ.

A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcr
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Bcr
Gene Accession No. P11274, Q6PAJ1
Gene Alias 5133400C09Rik; ABL; AI561783; AI853148; ALL; BCR; BCR activator of RhoGEF and GTPase; BCR protein; BCR, RhoGEF and GTPase activating protein; bcr/abl; BCR/FGFR1 chimera protein; BCR1; breakpoint cluster region; breakpoint cluster region homolog; breakpoint cluster region protein; c-ABL; c-abl oncogene 1; CML; D22S11; D22S662; EC 2.7.11.1; FGFR1/BCR chimera protein; JTK7; Kiaa3017; mKIAA3017; p150; PHL; renal carcinoma antigen NY-REN-26; RP11-83J21.1; v-abl
Gene Symbols Bcr
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human BCR (NP_004318.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110279, 309696, 613
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.