Learn More
BD Bacto™ Dehydrated Culture Media: Todd Hewitt Broth
Product Code. 11798952
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
For Use With (Application):
Hemolytic Streptococci growth
Form:
Dehydrated
Unit Size:
10kg
2kg
500g
Product Code. 11798952
Brand: BD 249220
For cultivation of group A streptococci, pathogenic microorganisms or as a blood culture medium
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Refrigerate
Form Dehydrated
Media Type Todd Hewitt Broth
Product Type Microbiology Media
Formulation Heart, infusion 3.1g/L, Neopeptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L
For Use With (Application) Hemolytic Streptococci growth
Packaging Type Pail
Quantity 10 kg