missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Phosphate Buffered Saline
Fluorescent Antibody reagents
Brand: BD 223143
Additional Details : Weight : 0.80000kg
Specifications
Phosphate Buffered Saline | |
Fluorescent Antibody reagents | |
10g | |
ISO/DIS, 11014 |
<30°C | |
1825 Days | |
Dehydrated | |
Vial |