missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Broth

Product Code. 11987245
Change view
Click to view available options
Quantity:
5 mL, 10/Pk.
5mL, Carton of 100 Size K Tubes
Media Type:
Brain Heart Infusion Broth
Unit Size:
Pack of 10
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Media Type unitSize
11987245 5mL, Carton of 100 Size K Tubes Brain Heart Infusion Broth Pack of 100
16678563 5 mL, 10/Pk. Brain Heart Infusion Broth Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11987245 Supplier BD Supplier No. 221813

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

General-purpose liquid media used in the cultivation of fastidious and nonfastidious microorganisms, including aerobic and anaerobic bacteria, from a variety of clinical and nonclinical materials

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Color Clear
Form Liquid
Manufacturing Origin United States
Media Type Brain Heart Infusion Broth
Formulation Brain heart infusion from (solids) 6g/L, Peptic digest of Animal Tissue 6g/L, Sodium Chloride 5g/L, Dextrose 3g/L, Pancreatic digest of Gelatin 14.5g/L, Disodium Phosphate 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms
Packaging Type K Tube
Quantity 5mL, Carton of 100 Size K Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.