Learn More
BD BBL™ Mycoflask™ Simmons Citrate Agar Prepared Media Slants
Click to view available options
:
Simmons citrate agar slants; Quantity 10
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | CE, AOAC, BAM, CCAM, COMPF, ISO |
| Color | Green |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Simmons Citrate Agar |
| Formulation | Ammonium Dihydrogen Phosphate 1g/L, Dipotassium Phosphate 1g/L, Sodium Chloride 5g/L, Sodium Citrate 2g/L, Magnesium Sulfate 0.2g/L, Agar 15g/L, Bromthymol blue 0.08g/L |
| Packaging Type | Tube with Tight Cap |
| Quantity | 10/Pk. |
For in vitro diagnostic use