Learn More
BD BBL™ Mycoflask™ TSI Agar™ Slants (Triple Sugar Iron Agar) Prepared Media
Product Code. 17382311
Click to view available options
Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
Product Code. 17382311
Brand: BD 221038
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, MCM9, SMD, SMWW, USDA
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type TSI Agar (Triple Sugar Iron)
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Sodium Chloride 5g/L, Lactose 10g/L, Sucrose 10g/L, Dextrose 1g/L, Ferrous Ammonium Sulfate 0.2g/L, Sodium Thiosulfate 0.2g/L, Phenol red 0.025g/L, Agar 13g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from variety of clinical and nonclinical specimens
Packaging Type Size K tubes with tight cap
Quantity 10/Pk.
For in vitro diagnostic use.