missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Replacement Reagents for Gram Stain Kits

For BBL Gram Stain Kits

Brand:  BD 212543

Additional Details : Weight : 3.80000kg


 View more versions of this product

Product Code. 11768213

  • € 350.00 / 3.80 L
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
  • Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles

Specifications

Specifications

Gram Iodine (Stabilized)
CE
15° to 30°C
3.8L
Used to stain microorganisms from cultures or specimens by the differential gram method
Iodine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P352-Wash with plenty of water/ soap.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : polyvinylpyrrolidone-iodine, potassium iodide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers