Learn More
BD BBL™ Replacement Reagents for Gram Stain Kits
Product Code. 11778213
Change view
Click to view available options
Quantity:
3.8 L
4 x 250 mL
Type:
Basic Fuchsin
Gram Iodine
Gram Iodine (Stabilized)
Safranin
Stain Type:
Basic Fuchsin
Iodine
Safranin
Unit Size:
1000mL
3.8 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Type Stain Type unitSize
11778213 4 x 250 mL Basic Fuchsin Basic Fuchsin 1000mL
11728203 4 x 250 mL Safranin Safranin 1000mL
11728213 3.8 L Safranin Safranin 3.8 L
11758213 4 x 250 mL Gram Iodine Iodine 1000mL
11768213 3.8 L Gram Iodine (Stabilized) Iodine 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11778213 Supplier BD Supplier No. 212544
For BBL Gram Stain Kits
  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
  • Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California
TRUSTED_SUSTAINABILITY
Specifications
Type Basic Fuchsin
Quantity 4 x 250 mL
Certifications/Compliance CE
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Storage Requirements 15° to 30°C
Stain Type Basic Fuchsin