missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Fosfomycin82

Product Code. p-4638830
Click to view available options
Quantity:
Each
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11727594

Brand: BD 231709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Fosfomycin
Concentration 200 μg
Packaging Type Disc
Quantity Each
compliance-icons
Product Identifier
  • BD BBL Sensi-Disc Fosfomycin 200
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : (2R-cis)-(3-Methyloxiranyl)phosphonic acid, compound with 2-amino-2-(hydroxymethyl)propane-1,3-diol

For in vitro diagnostic use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.