Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Norfloxacin54
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand:  BD 231647
Additional Details : Weight : 0.00940kg
Product Code. 17382391
  • € 185.00 / Pack of 10
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
Norfloxacin54
10μg
Disc
10 Pack
Certificates
Special Offers
Special Offers
For in vitro diagnostic use