Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Norfloxacin54
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand: BD 231647
Additional Details : Weight : 0.00940kg
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Norfloxacin54 | |
10μg | |
Disc | |
10 Pack |
For in vitro diagnostic use