Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Novobiocin (5μg)
Product Code. 17302381
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 17302381
Brand: BD 231314
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Novobiocin
Concentration 5 μg
Packaging Type Disc
Quantity 10/Pk.