Learn More
BD BBL™ Taxo™ A Discs (Vial)
Click to view available options
Packaging Type:
6 Vials
Single Vial
Quantity:
50 Discs
6 x 50 Discs
Unit Size:
Pack of 300
Pack of 50
Description
- Impregnated with approximately 0.04 units of bacitracin per disc
Taxo A discs are not for susceptibility testing.
Order Info
Vials of 50 discs
Specifications
Specifications
| Antimicrobial Agents | Bacitracin |
| Concentration | 0.04U |
| Content And Storage | -20°C to +8°C |
| For Use With (Application) | For differentiating Group A Streptococci |
| Packaging Type | 6 Vials |
| Quantity | 6 x 50 Discs |
For in vitro Diagnostic Use.