Learn More
BD Cytometric Bead Array (CBA) Human CD62L (L-Selectin); Flex Set, Bead D8
Description
Includes
Human CD62L (L-Selectin); Standard; Human CD62L (L-Selectin); PE Detection Reagent; Human CD62L (L-Selectin); Capture Bead D8
The BD CBA Human CD62L (L-Selectin) Flex Set is a bead-based immunoassay capable of measuring human soluble CD62L (L-Selectin) in serum, plasma and cell culture supernatant samples
- Reactivity: QC Tested: Human
- Bead Position: D8
- Limit of detection: 28.6pg/mL
With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), flow cytometer, and FCAP Array™ software
Specifications
Specifications
| Antigen | CD62L |
| Cross Reactivity | Human |
| Includes | Human CD62L (L-Selectin), standard, Human CD62L (L-Selectin), PE detection reagent, Human CD62L (L-Selectin), capture bead D8 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human CD62L (L-Selectin) flex set (bead D8) |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.