Learn More
BD Cytometric Bead Array (CBA) Human CD62P (P-Selectin); Flex Set, Bead D7
Bead-based immunoassay capable of measuring human soluble CD62P (P-Selectin) in serum, plasma and cell culture supernatant samples
Brand: BD 560426
Additional Details : Weight : 0.03600kg
Includes: Human CD62P (P-Selectin); Standard; Human CD62P (P-Selectin); PE Detection Reagent; Human CD62P (P-Selectin); Capture Bead D7
Description
Human reactivity was determined by testing samples with the BD CBA Human Soluble CD62P (P-Selectin) Flex Set.
- Format: Purified
Specifications
CD62P (P-Selectin) | |
Capture bead, detection reagent and standard | |
Flow Cytometry |
Human | |
100 Tests | |
Human CD62P (P-Selectin) flex set (bead D7) |