Learn More
BD Cytometric Bead Array (CBA) Human Enhanced Sensitivity Detection Reagent (Part B)
Description
Detection reagents used in BD™ CBA Human Enhanced Sensitivity Flex Set System consist of two parts, Analyte Specific Detection Reagent (Part A) and Enhanced Sensitivity Detection Reagent (Part B). Enhanced Sensitivity Detection Reagent (Part B) must be used in conjunction with Flex Kit, Master Buffer Kit, flow cytometer and FCAP Array™ Software.
- Provided as lyophilized cake
- Should not be used with Flex Sets from any other BD CBA System such as soluble protein, cell signalling etc
- For QC testing
- Reactivity: human
- Available in vial
Flow Cytometry
Specifications
Specifications
| Cross Reactivity | Human, Mouse |
| Quantity | 1 Vial |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry, Immunoassay |
| Product Type | Enhanced sensitivity detection reagent (Part B) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.