missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human IFNγ; Flex Set, Bead B8
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human IFNγ; Standard; Human IFNγ; PE Detection Reagent; Human IFNγ; Capture Bead B8
- Capable of measuring human interferon-γ (IFN-γ) in serum, plasma, and cell culture supernatant samples
Specifications
Specifications
| Antigen | IFN-gamma |
| Cross Reactivity | Human |
| Includes | Human IFNg, standard, Human IFNg, PE detection reagent, Human IFNg, capture bead B8 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IFN-gamma flex set (bead B8) |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction