Learn More
BD Cytometric Bead Array (CBA) Human IL-1α; Flex Set, Bead D6
Description
Includes
- Human IL-1α, Standard
- Human IL-1α PE Detection Reagent
- Human IL-1α; Capture Bead D6
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. The Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 2,500pg/mL.
- Human reactivity was determined by testing samples with CBA Human IL-1α Flex Set
- Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
Specifications
Specifications
| Antigen | IL-1alpha |
| Cross Reactivity | Human |
| Includes | Human IL-1a, standard, Human IL-1a PE detection reagent, Human IL-1a, capture bead D6 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-1alpha flex set (bead D6) |