Learn More
BD Cytometric Bead Array (CBA) Human IL-6 Enhanced Sensitivity Flex Set (Bead A7)
Measures human Interleukin-6 (IL-6) in serum, plasma and cell culture supernatant samples
Brand: BD 561512
Additional Details : Weight : 0.03600kg
Alert:
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Includes: Kit contents:
Enhanced Sensitivity Capture Bead A7: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150
Description
This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.
- Bead-based immunoassay
- Reactivity: human
- Standard assay range: 274 to 200000fg/mL
Applications:
Flow Cytometry/Immunoassay
Specifications
IL-6 | |
1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) | |
Serum, Plasma, Cell Culture Supernatant | |
Measures Human interleukin-6, flow cytometry and immunoassay |
Human | |
100 Tests | |
4°C | |
Human IL-6 enhanced sensitivity flex set (bead A7) |