Learn More
BD Difco™ Culture Media Additives: Penase Concentrate
Product Code. 15138763
Change view
Click to view available options
Quantity:
10 x 20 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15138763 10 x 20 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15138763 Supplier BD Supplier No. 215332
To enrich media for cultivating and isolating large variety of microorganisms and tissue cells BD Difco™ Culture Media Additives: Penase Concentrate is used with materials containing penicillin.
Penase and Penase Concentrate
  • For sterility testing and microbial count determination of materials containing penicillin
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Additive Penase
Quantity 10 x 20 mL