Learn More
BD Difco™ Culture Media Additives: Penase
Product Code. 15402414
Change view
Click to view available options
:
Penase; 10 x 20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15402414 Penase; 10 x 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15402414 Supplier BD Supplier No. 215331
To enrich media for cultivating and isolating large variety of microorganisms and tissue cells
Penase and Penase Concentrate
  • For sterility testing and microbial count determination of materials containing penicillin
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Additive Penase