Learn More
BD Difco™ Dehydrated Culture Media: Asparagine
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | <30°C |
| Additive | Asparagine |
| Form | Powder |
| For Use With (Application) | Chemical and microbiological usage |
| Packaging Type | Bottle |
| Quantity | 500 g |