missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Azide Blood Agar Base
Click to view available options
:
Azide Blood Agar Base; 500g
Description
- For use with blood in determining hemolytic reactions
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Azide Blood Agar Base |
| Product Type | Agar |
| Formulation | Proteose Peptone No. 3 4g/L, Pancreatic digest of Casein 5.8g/L, Beef extract 3g/L, Sodium Chloride 5g/L, Sodium Azide 0.2g/L, Agar 15g/L |
| For Use With (Application) | For isolating Streptococci and Staphylococci and, supplemented with Blood |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Product Identifier
- Bottle Azide Blood Agar Base 500G
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sodium Azide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction