Learn More
BD Difco™ Dehydrated Culture Media: Biotin Assay Medium
Click to view available options
:
Biotin Assay Medium; 100g
Description
Certifications
ISO, 9000
Specifications
Specifications
| Certifications/Compliance | ISO, 9000 |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Biotin Assay Medium |
| Product Type | Biotin Assay Medium |
| Formulation | Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Acetate 20/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 2mg/L, Riboflavin 2mg/L, Niacin 2mg/L, Calcium Pantothenate 2mg/L, Pyridoxine Hydrochloride 4mg/L, p-Aminobenzoic Acid 200μg/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L |
| For Use With (Application) | Isolation of Lactobacillus plantarum 8014 |
| Packaging Type | Bottle |
| Quantity | 100 g |