missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Eugon Agar

Product Code. 11749002
Click to view available options
:
Eugon Agar; 500g
This item is not returnable. View return policy

Product Code. 11749002

Brand: BD 258910

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

General-purpose medium for cultivating a wide variety of microorganisms

  • Formulation: Proteose Peptone 7.5g/L, Pancreatic digest of Casein 7.5g/L, Soy Peptone 5g/L, Dextrose 5.5g/L, L-Cystine 0.7g/L, Sodium Chloride 4g/L, Sodium Sulfite 0.2g/L, Agar 15g/L

Certifications

CE, COMPF

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Eugon Agar
Product Type Agar
Formulation Proteose Peptone 7.5g/L, Pancreatic digest of Casein 7.5g/L, Soy Peptone 5g/L, Dextrose 5.5g/L, L-Cystine 0.7g/L, Sodium Chloride 4g/L, Sodium Sulfite 0.2g/L, Agar 15g/L
For Use With (Application) Cultivation of a variety of organisms, well suited for large scale vaccine production
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.