missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: King Medium A (Pseudomonas agar P)

Product Code. 11798902
Click to view available options
:
King Medium A (Pseudomonas Agar P); 500g
This item is not returnable. View return policy

Product Code. 11798902

Brand: BD 244910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with Glycerol for detection and differentiation of P. aeruginosa based on pyocyanin production

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Content And Storage Refrigerate
Form Dehydrated
Media Type King Medium A
Product Type Microbiology Media
Formulation Pancreatic digest of Gelatin 20g/L, Magnesium Chloride 1.4g/L, Potassium Sulfate 10g/L, Agar 15g/L
For Use With (Application) Used with Glycerol for detection and differentiation of P. aeruginosa based on Pyocyanin production
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.