missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Liver Infusion Broth
Click to view available options
:
Liver Infusion Broth; 500g
Description
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Liver Infusion Broth |
| Product Type | Microbiology Media |
| Formulation | Beef liver infusion from 500g 20g/L, Proteose Peptone 10g/L, Sodium Chloride 5g/L |
| For Use With (Application) | Cultivation of variety of organisms, particularly Brucella and anaerobes |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction