missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Lysine Iron Agar
Click to view available options
:
Lysine Iron Agar; 500g
Description
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, CCAM, CMPH2, COMPF, MCM9, SMD, SMWW, USDA |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Lysine Iron Agar |
| Product Type | Agar |
| Formulation | Peptone 5g/L, Yeast extract 3g/L, Dextrose 1g/L, L-Lysine HCl 10g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Thiosulfate 0.04g/L, Bromcresol Purple 0.02g/L, Agar 15g/L |
| For Use With (Application) | Differential medium to detect organisms that decarboxylate or deaminate Lysine and produce Hydrogen sulfide |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction